siRNA Details
Gene Symbol: Hoxb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047091.1 | NP_001040556.1 | 438 | |||
GenBank mRNA | DQ399532.1 | - | 438 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hoxb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047091.1 | NP_001040556.1 | 438 | |||
GenBank mRNA | DQ399532.1 | - | 438 |