siRNA Details
| Gene Symbol: Msl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107048.1 | NP_001100518.1 | 2 | 928 | ||
| XM_006247304.2 | XP_006247366.1 | 2 | 1247 | |||
| GenBank mRNA | FQ228290.1 | - | 2 | 882 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msl1| Gene Symbol: Msl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107048.1 | NP_001100518.1 | 2 | 928 | ||
| XM_006247304.2 | XP_006247366.1 | 2 | 1247 | |||
| GenBank mRNA | FQ228290.1 | - | 2 | 882 | ||