siRNA Details
| Gene Symbol: Msl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107048.1 | NP_001100518.1 | 1, 2 | 813 | ||
| XM_006247304.2 | XP_006247366.1 | 1, 2 | 1132 | |||
| GenBank mRNA | FQ228290.1 | - | 1, 2 | 767 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_028722, BC058629, BC043039, BC055715, AK208034, and others.

Msl1
Genomic Map