siRNA Details
| Gene Symbol: Rapgefl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768202.1 | XP_008766424.1 | 4 | 1160 | ||
| XM_008768203.1 | XP_008766425.1 | 4 | 1161 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rapgefl1| Gene Symbol: Rapgefl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768202.1 | XP_008766424.1 | 4 | 1160 | ||
| XM_008768203.1 | XP_008766425.1 | 4 | 1161 | |||