siRNA Details
| Gene Symbol: Smarce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024993.1 | NP_001020164.1 | 6 | 417 | ||
| XM_008768051.1 | XP_008766273.1 | 6 | 759 | |||
| GenBank mRNA | BC091314.1 | - | 6 | 473 | 7312354 | |
| BC092210.1 | - | 6 | 417 | 7311688 | ||
| EU327027.1 | - | 6 | 422 | |||
| EU327028.1 | - | 5 | 378 | |||
| EU327029.1 | - | 6 | 475 | |||
| EU327030.1 | - | 5 | 339 | |||
| EU327031.1 | - | 4 | 326 | |||
| FQ213000.1 | - | 6 | 405 | |||

Smarce1
Genomic Map