siRNA Details
| Gene Symbol: Smarce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024993.1 | NP_001020164.1 | 7 | 690 | ||
| XM_008768051.1 | XP_008766273.1 | 7 | 1032 | |||
| GenBank mRNA | BC091314.1 | - | 7 | 746 | 7312354 | |
| BC092210.1 | - | 7 | 690 | 7311688 | ||
| FQ213000.1 | - | 7 | 678 | |||
| Gene Symbol: LOC103694314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772543.1 | XP_008770765.1 | 1 | 2565 | ||

Smarce1
Genomic Map