siRNA Details
| Gene Symbol: Zfp385c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247466.2 | XP_006247528.1 | 6 | 2301 | ||
| XM_008768216.1 | XP_008766438.1 | 7 | 1024 | |||
| XM_008768217.1 | XP_008766439.1 | 6 | 2301 | |||
| XM_008768218.1 | XP_008766440.1 | 6 | 841 | |||
| GenBank mRNA | BC162044.1 | - | 5 | 749 | ||

Zfp385c
Genomic Map