siRNA Details
| Gene Symbol: Dhx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098788.1 | NP_001092258.1 | 5 | 723 | ||
| XM_006247314.2 | XP_006247376.1 | 6 | 1292 | |||
| XM_006247315.2 | XP_006247377.1 | 6 | 1281 | |||
| GenBank mRNA | BP488074.1 | - | 3 | 196 | ||

Dhx58
Genomic Map