siRNA Details
| Gene Symbol: Rnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010953.1 | NP_001010953.1 | 4 | 548 | ||
| GenBank mRNA | BC089058.1 | - | 4 | 548 | 7315480 | |
| D82867.1 | - | 4 | 578 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnd2| Gene Symbol: Rnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010953.1 | NP_001010953.1 | 4 | 548 | ||
| GenBank mRNA | BC089058.1 | - | 4 | 548 | 7315480 | |
| D82867.1 | - | 4 | 578 | |||