siRNA Details
| Gene Symbol: Arl4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107052.1 | NP_001100522.1 | 2 | 531 | ||
| XM_008768053.1 | XP_008766275.1 | 2 | 1517 | |||
| XM_008768054.1 | XP_008766276.1 | 3 | 1377 | |||
| GenBank mRNA | BC167047.1 | - | 2 | 506 | 8367403 | |
| EF688597.1 | - | 2 | 506 | |||

Arl4d
Genomic Map