siRNA Details
| Gene Symbol: Nags | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107053.1 | NP_001100523.1 | 4 | 1005 | ||
| XM_006247328.2 | XP_006247390.1 | 5 | 1825 | |||
| XM_008768055.1 | XP_008766277.1 | 5 | 1825 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nags| Gene Symbol: Nags | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107053.1 | NP_001100523.1 | 4 | 1005 | ||
| XM_006247328.2 | XP_006247390.1 | 5 | 1825 | |||
| XM_008768055.1 | XP_008766277.1 | 5 | 1825 | |||