siRNA Details
| Gene Symbol: Tmub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031651.2 | NP_001026821.1 | 3 | 1332 | ||
| XM_006247331.2 | XP_006247393.1 | 3 | 989 | |||
| XM_006247332.2 | XP_006247394.1 | 4 | 987 | |||
| GenBank mRNA | BC085794.1 | - | 3 | 1332 | 7109581 | |
| BC099074.1 | - | 3 | 1333 | 7109079 | ||

Tmub2
Genomic Map