siRNA Details
| Gene Symbol: Acbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012013.1 | NP_001012013.1 | 4, 5 | 576 | ||
| XM_006247508.2 | XP_006247570.1 | 5, 6 | 564 | |||
| XM_006247509.2 | XP_006247571.1 | 5, 6 | 566 | |||
| XM_006247510.2 | XP_006247572.1 | 5, 6 | 565 | |||
| XM_006247511.2 | XP_006247573.1 | 5, 6 | 564 | |||
| XM_006247512.2 | XP_006247574.1 | 5, 6 | 565 | |||
| XM_008768284.1 | XP_008766506.1 | 6, 7 | 667 | |||
| XM_008768285.1 | XP_008766507.1 | 5, 6 | 555 | |||
| XM_008768286.1 | XP_008766508.1 | 5, 6 | 566 | |||
| XM_008768287.1 | XP_008766509.1 | 5, 6 | 565 | |||
| GenBank mRNA | BC076387.1 | - | 4, 5 | 576 | 7109482 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025988, AK148438, AK008243, AK079604, BC048371, and others.

Acbd4
Genomic Map