siRNA Details
| Gene Symbol: Plekhm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009677.1 | NP_001009677.1 | 5 | 1128 | ||
| XM_006247527.2 | XP_006247589.1 | 5 | 1168 | |||
| XM_008768292.1 | XP_008766514.1 | 4 | 948 | |||
| GenBank mRNA | BC087058.1 | - | 5 | 1128 | 7124252 | |

Plekhm1
Genomic Map