siRNA Details
| Gene Symbol: March10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013973.1 | NP_001013995.1 | 3 | 329 | ||
| XM_006247579.2 | XP_006247641.1 | 3 | 497 | |||
| XM_006247580.2 | XP_006247642.1 | 4 | 616 | |||
| XM_006247585.2 | XP_006247647.1 | 3 | 474 | |||
| GenBank mRNA | AB552845.1 | - | 1 | 74 | ||
| AB552846.1 | - | 1 | 74 | |||
| BC083567.1 | - | 3 | 329 | 7112851 | ||

March10
Genomic Map