siRNA Details
| Gene Symbol: Cyb561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107056.2 | NP_001100526.1 | 5 | 580 | ||
| XM_008768357.1 | XP_008766579.1 | 4 | 382 | |||
| GenBank mRNA | BC169069.1 | - | 5 | 580 | 5623837 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyb561| Gene Symbol: Cyb561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107056.2 | NP_001100526.1 | 5 | 580 | ||
| XM_008768357.1 | XP_008766579.1 | 4 | 382 | |||
| GenBank mRNA | BC169069.1 | - | 5 | 580 | 5623837 | |