siRNA Details
| Gene Symbol: Ddx42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107059.1 | NP_001100529.1 | 5, 6 | 723 | ||
| XM_006247596.2 | XP_006247658.1 | 5, 6 | 677 | |||
| XM_006247597.2 | XP_006247659.1 | 4, 5 | 488 | |||
| GenBank mRNA | FQ221897.1 | - | 4, 5 | 288 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_028074, AK171730, AK049311, AK209812, AK169816, and others.

Ddx42
Genomic Map