siRNA Details
| Gene Symbol: Tex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247645.1 | XP_006247707.1 | 10 | 3103 | ||
| XM_006247646.2 | XP_006247708.1 | 10 | 3935 | |||
| XM_006247647.1 | XP_006247709.1 | 10 | 3097 | |||
| GenBank mRNA | BC083806.1 | - | 9 | 1487 | ||
| BC097967.1 | - | 6 | 765 | |||

Tex2
Genomic Map