siRNA Details
| Gene Symbol: Smurf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107061.1 | NP_001100531.1 | 13 | 1362 | ||
| XM_008768365.1 | XP_008766587.1 | 12 | 1267 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smurf2| Gene Symbol: Smurf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107061.1 | NP_001100531.1 | 13 | 1362 | ||
| XM_008768365.1 | XP_008766587.1 | 12 | 1267 | |||