siRNA Details
| Gene Symbol: Wipi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127297.1 | NP_001120769.1 | 10 | 1105 | ||
| XM_008768366.1 | XP_008766588.1 | 10 | 1724 | |||
| XM_008768367.1 | XP_008766589.1 | 10 | 1002 | |||
| GenBank mRNA | BC166829.1 | - | 10 | 1105 | 8373479 | |

Wipi1
Genomic Map