siRNA Details
| Gene Symbol: Btbd17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134534.1 | NP_001128006.1 | 3 | 592 | ||
| XM_006247691.2 | XP_006247753.1 | 3 | 617 | |||
| XM_006247692.2 | XP_006247754.1 | 3 | 1388 | |||
| XM_008768371.1 | XP_008766593.1 | 2 | 1047 | |||

Btbd17
Genomic Map