siRNA Details
| Gene Symbol: Caskin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107065.1 | NP_001100535.1 | 2 | 616 | ||
| XM_006247717.2 | XP_006247779.1 | 2 | 654 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Caskin2| Gene Symbol: Caskin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107065.1 | NP_001100535.1 | 2 | 616 | ||
| XM_006247717.2 | XP_006247779.1 | 2 | 654 | |||