siRNA Details
| Gene Symbol: Mrpl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009369.2 | NP_001009369.2 | 3 | 309 | ||
| XM_008768372.1 | XP_008766594.1 | 3 | 1097 | |||
| GenBank mRNA | BC087096.1 | - | 3 | 306 | 7191805 | |
| CO567099.1 | - | 3 | 309 | |||

Mrpl38
Genomic Map