siRNA Details
| Gene Symbol: Tbc1d16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247938.2 | XP_006248000.1 | 6 | 1586 | ||
| XM_008768486.1 | XP_008766708.1 | 7 | 3884 | |||
| XM_221188.8 | XP_221188.5 | 6 | 1583 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d16| Gene Symbol: Tbc1d16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247938.2 | XP_006248000.1 | 6 | 1586 | ||
| XM_008768486.1 | XP_008766708.1 | 7 | 3884 | |||
| XM_221188.8 | XP_221188.5 | 6 | 1583 | |||