siRNA Details
| Gene Symbol: Slc38a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727846.4 | XP_002727892.2 | 17 | 3154 | ||
| XM_006247943.2 | XP_006248005.1 | 17 | 3151 | |||
| XM_006247944.2 | XP_006248006.1 | 17 | 3152 | |||
| XM_006247945.2 | XP_006248007.1 | 16 | 3127 | |||
| XM_006247946.2 | XP_006248008.1 | 16 | 3127 | |||
| XM_221195.8 | XP_221195.5 | 16 | 3130 | |||
| GenBank mRNA | BC098794.1 | - | 1 | 202 | ||

Slc38a10
Genomic Map