siRNA Details
| Gene Symbol: Notum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013975.2 | NP_001013997.2 | 9 | 1009 | ||
| XM_006247887.2 | XP_006247949.1 | 9 | 1242 | |||
| XM_006247888.2 | XP_006247950.1 | 8 | 1579 | |||
| GenBank mRNA | BC090016.1 | - | 2 | 391 | ||
| CO562343.1 | - | 4 | 342 | |||

Notum
Genomic Map