siRNA Details
| Gene Symbol: Mrpl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029900.1 | NP_001025071.1 | 4 | 599 | ||
| GenBank mRNA | FQ228179.1 | - | 4 | 413 | ||
| FQ233456.1 | - | 4 | 415 | |||
| FQ234892.1 | - | 4 | 413 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl12| Gene Symbol: Mrpl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029900.1 | NP_001025071.1 | 4 | 599 | ||
| GenBank mRNA | FQ228179.1 | - | 4 | 413 | ||
| FQ233456.1 | - | 4 | 415 | |||
| FQ234892.1 | - | 4 | 413 | |||