siRNA Details
| Gene Symbol: Cd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107074.1 | NP_001100544.1 | 2 | 250 | ||
| GenBank mRNA | BC167030.1 | - | 2 | 222 | 7378211 | |
| FQ226313.1 | - | 2 | 252 | |||
| FQ226643.1 | - | 2 | 252 | |||
| FQ227263.1 | - | 2 | 252 | |||

Cd7
Genomic Map