siRNA Details
| Gene Symbol: Ogfod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013976.1 | NP_001013998.1 | 5, 6 | 606 | ||
| GenBank mRNA | BC088238.1 | - | 5, 6 | 606 | 7326036 | |
| FQ225392.1 | - | 5, 6 | 595 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ogfod3| Gene Symbol: Ogfod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013976.1 | NP_001013998.1 | 5, 6 | 606 | ||
| GenBank mRNA | BC088238.1 | - | 5, 6 | 606 | 7326036 | |
| FQ225392.1 | - | 5, 6 | 595 | |||