siRNA Details
| Gene Symbol: Dgcr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107080.2 | NP_001100550.1 | 5 | 546 | ||
| XM_006248609.2 | XP_006248671.1 | 5 | 584 | |||
| GenBank mRNA | BC158828.1 | - | 5 | 546 | 7378165 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dgcr6| Gene Symbol: Dgcr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107080.2 | NP_001100550.1 | 5 | 546 | ||
| XM_006248609.2 | XP_006248671.1 | 5 | 584 | |||
| GenBank mRNA | BC158828.1 | - | 5 | 546 | 7378165 | |