siRNA Details
| Gene Symbol: Map3k13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013978.2 | NP_001014000.2 | 591 | |||
| GenBank mRNA | BC081976.1 | - | 472 | 7127060 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k13| Gene Symbol: Map3k13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013978.2 | NP_001014000.2 | 591 | |||
| GenBank mRNA | BC081976.1 | - | 472 | 7127060 | ||