siRNA Details
Gene Symbol: Map3k13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013978.2 | NP_001014000.2 | 1715 | |||
GenBank mRNA | BC081976.1 | - | 1596 | 7127060 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Map3k13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013978.2 | NP_001014000.2 | 1715 | |||
GenBank mRNA | BC081976.1 | - | 1596 | 7127060 |