siRNA Details
| Gene Symbol: Slc51a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107087.1 | NP_001100557.1 | 3 | 567 | ||
| XM_008768725.1 | XP_008766947.1 | 3 | 388 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc51a| Gene Symbol: Slc51a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107087.1 | NP_001100557.1 | 3 | 567 | ||
| XM_008768725.1 | XP_008766947.1 | 3 | 388 | |||