siRNA Details
| Gene Symbol: Tnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008336.1 | NP_001008337.1 | 2 | 384 | ||
| GenBank mRNA | BC085825.1 | - | 2 | 384 | 7130376 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnk2| Gene Symbol: Tnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008336.1 | NP_001008337.1 | 2 | 384 | ||
| GenBank mRNA | BC085825.1 | - | 2 | 384 | 7130376 | |