siRNA Details
| Gene Symbol: Muc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248471.2 | XP_006248533.1 | 3 | 3408 | ||
| XM_008768775.1 | XP_008766997.1 | 3 | 3048 | |||
| XM_008768776.1 | XP_008766998.1 | 3 | 2316 | |||
| XM_008768777.1 | XP_008766999.1 | 3 | 1585 | |||
| GenBank mRNA | U06752.1 | - | 3 | 1882 | ||

Muc4
Genomic Map