siRNA Details
| Gene Symbol: Osbpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107090.2 | NP_001100560.1 | 11, 12 | 2490 | ||
| XM_006248450.2 | XP_006248512.1 | 10, 11 | 2545 | |||
| XM_006248451.2 | XP_006248513.1 | 11, 12 | 2121 | |||
| XM_006248452.2 | XP_006248514.1 | 11, 12 | 2002 | |||
| XM_008768726.1 | XP_008766948.1 | 11, 12 | 2271 | |||
| GenBank mRNA | BC168952.1 | - | 11, 12 | 2490 | 7453666 | |

Osbpl11
Genomic Map