siRNA Details
Gene Symbol: Dirc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012017.1 | NP_001012017.1 | 2 | 667 | ||
GenBank mRNA | BC081960.1 | - | 2 | 667 | 7124589 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dirc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012017.1 | NP_001012017.1 | 2 | 667 | ||
GenBank mRNA | BC081960.1 | - | 2 | 667 | 7124589 |