siRNA Details
| Gene Symbol: Parp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103351.2 | NP_001096821.1 | 5 | 1224 | ||
| XM_006248411.1 | XP_006248473.1 | 5 | 1050 | |||
| XM_008768727.1 | XP_008766949.1 | 5 | 2333 | |||
| XM_008768728.1 | XP_008766950.1 | 4 | 2978 | |||
| XM_008768729.1 | XP_008766951.1 | 5 | 1154 | |||
| GenBank mRNA | BC128756.1 | - | 5 | 1227 | 7105391 | |

Parp9
Genomic Map