siRNA Details
| Gene Symbol: Ildr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127536.3 | NP_001121008.2 | 3, 4 | 474 | ||
| XM_006248394.2 | XP_006248456.1 | 3, 4 | 543 | |||
| GenBank mRNA | BC166709.1 | - | 3, 4 | 474 | 7929589 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ildr1| Gene Symbol: Ildr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127536.3 | NP_001121008.2 | 3, 4 | 474 | ||
| XM_006248394.2 | XP_006248456.1 | 3, 4 | 543 | |||
| GenBank mRNA | BC166709.1 | - | 3, 4 | 474 | 7929589 | |