siRNA Details
| Gene Symbol: B4galt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012018.1 | NP_001012018.1 | 2 | 301 | ||
| GenBank mRNA | BC081866.1 | - | 2 | 301 | 7096554 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

B4galt4| Gene Symbol: B4galt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012018.1 | NP_001012018.1 | 2 | 301 | ||
| GenBank mRNA | BC081866.1 | - | 2 | 301 | 7096554 | |