siRNA Details
| Gene Symbol: RGD1311458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009678.1 | NP_001009678.1 | 8, 9 | 1279 | ||
| GenBank mRNA | BC088456.1 | - | 8, 9 | 1279 | 7127273 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311458| Gene Symbol: RGD1311458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009678.1 | NP_001009678.1 | 8, 9 | 1279 | ||
| GenBank mRNA | BC088456.1 | - | 8, 9 | 1279 | 7127273 | |