siRNA Details
| Gene Symbol: Abhd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123352.1 | NP_001116824.1 | 4, 5 | 555 | ||
| GenBank mRNA | BC088235.1 | - | 4, 5 | 548 | ||
| BC161919.1 | - | 4, 5 | 555 | 8369987 | ||
| Gene Symbol: LOC103693563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768673.1 | XP_008766895.1 | 4, 5 | 685 | ||
| XM_008768674.1 | XP_008766896.1 | 4, 5 | 1292 | |||

Abhd10
Genomic Map