siRNA Details
| Gene Symbol: Bbx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079938.1 | NP_001073407.1 | 9 | 1332 | ||
| XM_008768634.1 | XP_008766856.1 | 9 | 1224 | |||
| XM_008768635.1 | XP_008766857.1 | 9 | 1419 | |||
| XM_008768636.1 | XP_008766858.1 | 10 | 1533 | |||
| XM_008768637.1 | XP_008766859.1 | 10 | 1338 | |||
| XM_008768638.1 | XP_008766860.1 | 9 | 1224 | |||
| XM_008768639.1 | XP_008766861.1 | 9 | 1419 | |||
| XM_008768640.1 | XP_008766862.1 | 9 | 1110 | |||
| XM_008768641.1 | XP_008766863.1 | 9 | 1224 | |||
| GenBank mRNA | BC129113.1 | - | 9 | 1332 | 7112122 | |
| Gene Symbol: LOC100912484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751043.3 | XP_003751091.1 | 1 | 987 | ||

Bbx
Genomic Map