siRNA Details
| Gene Symbol: Igsf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006990.2 | NP_001006991.1 | 5 | 1129 | ||
| XM_006248155.2 | XP_006248217.1 | 5 | 994 | |||
| XM_006248156.2 | XP_006248218.1 | 4 | 693 | |||
| GenBank mRNA | AY439284.1 | - | 4 | 775 | ||
| BC086342.1 | - | 5 | 835 | |||
| BC097947.1 | - | 5 | 1129 | 7373302 | ||

Igsf5
Genomic Map