siRNA Details
| Gene Symbol: Zbtb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107097.1 | NP_001100567.1 | 5 | 1910 | ||
| XM_006248251.2 | XP_006248313.1 | 5 | 1586 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb11| Gene Symbol: Zbtb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107097.1 | NP_001100567.1 | 5 | 1910 | ||
| XM_006248251.2 | XP_006248313.1 | 5 | 1586 | |||