siRNA Details
Gene Symbol: LOC100911145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751038.3 | XP_003751086.1 | 13 | 1848 |
Gene Symbol: Gpr128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107098.1 | NP_001100568.1 | 2020 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
