siRNA Details
| Gene Symbol: Cpox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037095.1 | NP_001032172.1 | 3 | 833 | ||
| GenBank mRNA | BC107664.1 | - | 3 | 833 | 7936479 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpox| Gene Symbol: Cpox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037095.1 | NP_001032172.1 | 3 | 833 | ||
| GenBank mRNA | BC107664.1 | - | 3 | 833 | 7936479 | |