siRNA Details
| Gene Symbol: Olr1533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000496.1 | NP_001000496.1 | 1 | 511 | ||
| Gene Symbol: Olr1543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001104.1 | NP_001001104.1 | 1 | 511 | ||

Olr1533
Genomic Map