siRNA Details
| Gene Symbol: SHOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001269039.2 | NP_001255968.1 | 4 | 1223 | ||
| NM_001324336.1 | NP_001311265.1 | 5 | 1318 | |||
| NM_001324337.1 | NP_001311266.1 | 6 | 1546 | |||
| NM_007373.3 | NP_031399.2 | 5 | 1361 | |||
| NR_136749.1 | 4 | 424 | ||||
| XM_017016702.1 | XP_016872191.1 | 5 | 1266 | |||
| XM_017016703.1 | XP_016872192.1 | 5 | 7083 | |||
| XM_017016704.1 | XP_016872193.1 | 5 | 1265 | |||
| GenBank mRNA | AB020669.1 | - | 5 | 1289 | ||
| AF054828.1 | - | 5 | 1283 | |||
| AF068920.1 | - | 4 | 1012 | |||
| AK090820.1 | - | 3 | 382 | |||
| AK292833.1 | - | 6 | 1487 | |||
| AK293271.1 | - | 4 | 363 | |||
| BC044752.1 | - | 5 | 1297 | |||
| BC050445.1 | - | 4 | 1216 | 6067669 | ||

SHOC2
Genomic Map