siRNA Details
| Gene Symbol: Arl13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107101.1 | NP_001100571.1 | 2 | 380 | ||
| XM_006240716.2 | XP_006240778.1 | 1 | 161 | |||
| XM_006240717.2 | XP_006240779.1 | 1 | 159 | |||
| XM_006240718.2 | XP_006240780.1 | 1 | 159 | |||

Arl13b
Genomic Map